Gene Transcripts

Gene Symbol
MSTO1
Entrez Gene ID
55154
Gene Name
misato 1, mitochondrial distribution and morphology regulator
Gene Aliases
LST005, MST
Chromosome Location
Chr. 1: 155610170 - 155614967 on Build GRCh38
UniGene ID
Hs.656547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_046293.1 4 459
XM_006711423.2 XP_006711486.1 4 403
XM_011509707.1 XP_011508009.1 4 860
XM_017001603.1 XP_016857092.1 4 391
XM_017001605.1 XP_016857094.1 4 860
XM_017001609.1 XP_016857098.1 4 403
XM_017001611.1 XP_016857100.1 4 860
XM_017001612.1 XP_016857101.1 3 296
XM_017001614.1 XP_016857103.1 4 403
XM_017001615.1 XP_016857104.1 4 403
GenBank mRNA AK309573.1 - 4 380
BC070067.1 - 4 411
CR936872.1 - 2 1015

Gene Symbol
MSTO2P
Entrez Gene ID
100129405
Gene Name
misato family member 2, pseudogene
Gene Aliases
MSTO2
Chromosome Location
Chr. 1: 155745768 - 155750688 on Build GRCh38
UniGene ID
Hs.656361
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024117.2 4 441
GenBank mRNA AK295146.1 - 4 441
BC099909.1 - 4 391
DC387604.1 - 4 441

View Full Product Details