Gene Transcripts

Gene Symbol
NCRUPAR
Entrez Gene ID
100302746
Gene Name
non-protein coding RNA, upstream of F2R/PAR1
Gene Aliases
NCRNA00193, ncR-uPAR
Chromosome Location
Chr. 5: 76712403 - 76712888 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028375.2 1 405

View Full Product Details