Gene Transcripts

Gene Symbol
Rian
Entrez Gene ID
75745
Gene Name
RNA imprinted and accumulated in nucleus
Gene Aliases
5530401N18Rik, C130089L09Rik, Meg8
Chromosome Location
Chr. 12: 109603945 - 109661716 on Build GRCm38
UniGene ID
Mm.293263
Species
Mus musculus
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB063319.1 - LIT1623
Rian
2 2327

View Full Product Details