Gene Transcripts
- Gene Symbol
- HERC2P2
- Entrez Gene ID
- 400322
- Gene Name
- hect domain and RLD 2 pseudogene 2
- Gene Aliases
- D15F37S3, MN7
- Chromosome Location
- Chr. 15: 22494837 - 22590831 on Build GRCh38
- UniGene ID
- Hs.458334
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF041080.1 |
- |
LIT2190 HERC2P2 |
32 |
4947 |
|
View Full Product Details