Gene Transcripts

A Gene Symbol is not available because the non-coding siRNA was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID. Non-coding siRNAs are designed to RefSeq (NR_) or to curated sequences from RNAdb represented by GenBank RNA or ESTdb accessions.


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AA602136.3 - 1 85 Chr. 11 - 2640573 - 2640930 [+]
GenBankmRNA AJ006345.3 - 2 195708 Chr. 11 - 2444100 - 2849207 [+]

View Full Product Details