Gene Transcripts

Gene Symbol
STUM
Entrez Gene ID
375057
Gene Name
stum, mechanosensory transduction mediator homolog
Gene Aliases
C1orf95
Chromosome Location
Chr. 1: 226548238 - 226609214 on Build GRCh38
UniGene ID
Hs.116827
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003665.3 NP_001003665.1 4 5123
XM_011544183.2 XP_011542485.1 3 5676
GenBank mRNA AF035308.1 - 1 671

View Full Product Details