Gene Transcripts

Gene Symbol
KLHL36
Entrez Gene ID
79786
Gene Name
kelch like family member 36
Gene Aliases
C16orf44
Chromosome Location
Chr. 16: 84648504 - 84667686 on Build GRCh38
UniGene ID
Hs.222731
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303451.1 NP_001290380.1 4 6052
NM_024731.3 NP_079007.2 5 6241
XM_005256149.2 XP_005256206.1 5 6455
XM_017023674.1 XP_016879163.1 5 6203
XM_017023675.1 XP_016879164.1 5 6223
GenBank mRNA AK056197.1 - 1 1235
AK056428.1 - 1 3039
AK074056.1 - 2 5168


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK057887.1 - 1075 Chr. - - [-]

View Full Product Details