Gene Transcripts
- Gene Symbol
- REPS1
- Entrez Gene ID
- 85021
- Gene Name
- RALBP1 associated Eps domain containing 1
- Gene Aliases
- RALBP1
- Chromosome Location
- Chr. 6: 138904015 - 138988282 on Build GRCh38
- UniGene ID
- Hs.334603
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209143.1 |
- |
6 |
4304 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AL049259.1 |
- |
1 |
110 |
|
Chr. 6 - 138921578 - 138927002 [-] |
View Full Product Details