Gene Transcripts
- Gene Symbol
- FBXO7
- Entrez Gene ID
- 25793
- Gene Name
- F-box protein 7
- Gene Aliases
- FBX, FBX07, FBX7, PARK15, PKPS
- Chromosome Location
- Chr. 22: 32474682 - 32498831 on Build GRCh38
- UniGene ID
- Hs.5912
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC041004.1 |
- |
1 |
176 |
|
View Full Product Details