Gene Transcripts
- Gene Symbol
- TMEM87A
- Entrez Gene ID
- 25963
- Gene Name
- transmembrane protein 87A
- Gene Aliases
- Chromosome Location
- Chr. 15: 42210452 - 42273584 on Build GRCh38
- UniGene ID
- Hs.511138
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK023569.1 |
- |
1 |
1486 |
|
View Full Product Details