Gene Transcripts
- Gene Symbol
- PKI55
- Entrez Gene ID
- 150967
- Gene Name
- DKFZp434H1419
- Gene Aliases
- Chromosome Location
- Chr. 2: 216216889 - 216220192 on Build GRCh38
- UniGene ID
- Hs.56876
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037701.1 |
|
1 |
918 |
|
GenBank mRNA |
AK125369.1 |
- |
1 |
762 |
|
AL137534.1 |
- |
1 |
222 |
|
View Full Product Details