Gene Transcripts
- Gene Symbol
- LGALS3
- Entrez Gene ID
- 3958
- Gene Name
- lectin, galactoside binding soluble 3
- Gene Aliases
- CBP35, GAL3, GALBP, GALIG, L31, LGALS2, MAC2
- Chromosome Location
- Chr. 14: 55129217 - 55145430 on Build GRCh38
- UniGene ID
- Hs.531081
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209391.1 |
- |
1 |
1690 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK091776.1 |
- |
1 |
1848 |
|
Chr. 14 - 55132966 - 55135154 [+] |
View Full Product Details