Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001197026.1 | NP_001183955.1 | 14 | 3229 | |
XM_011515596.2 | XP_011513898.1 | 13 | 2745 | ||
XM_017012746.1 | XP_016868235.1 | 14 | 3236 | ||
XM_017012747.1 | XP_016868236.1 | 14 | 3269 | ||
GenBank mRNA | CR627312.1 | - | 1 | 969 |