Gene Transcripts

Gene Symbol
POU3F3
Entrez Gene ID
5455
Gene Name
POU class 3 homeobox 3
Gene Aliases
BRN1, OTF8, brain-1, oct-8
Chromosome Location
Chr. 2: 104854068 - 104858574 on Build GRCh38
UniGene ID
Hs.248158
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006236.2 NP_006227.1 1 609
GenBank mRNA CX873380.1 - 1 549


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AL534719.3 - 1 568 Chr. 2 - 104854109 - 104855155 [+]
GenBankmRNA BF929868.1 - 1 293 Chr. 2 - 104854395 - 104854868 [+]
GenBankmRNA AK125157.1 - 562 Chr. - - [-]
GenBankmRNA CN266660.1 - 1 155 Chr. 2 - 104854524 - 104855038 [+]

View Full Product Details