Gene Transcripts

Gene Symbol
FAM204A
Entrez Gene ID
63877
Gene Name
family with sequence similarity 204 member A
Gene Aliases
C10orf84, bA319I23.1
Chromosome Location
Chr. 10: 118305889 - 118342352 on Build GRCh38
UniGene ID
Hs.105575
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005270024.1 XP_005270081.1 10 5994
GenBank mRNA BX538256.1 - 1 2059


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BX107735.1 - 1 348 Chr. 10 - 118305887 - 118306356 [-]
GenBankmRNA BU785999.1 - 1 555 Chr. 10 - 118305942 - 118306562 [-]
GenBankmRNA AA479591.1 - 1 347 Chr. 10 - 118305891 - 118306356 [-]

View Full Product Details