Gene Transcripts
- Gene Symbol
- ZNRF1
- Entrez Gene ID
- 84937
- Gene Name
- zinc and ring finger 1, E3 ubiquitin protein ligase
- Gene Aliases
- NIN283
- Chromosome Location
- Chr. 16: 74999017 - 75110994 on Build GRCh38
- UniGene ID
- Hs.427284
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK128349.1 |
- |
1 |
77 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB150897.1 |
- |
1 |
77 |
|
Chr. 16 - 75104060 - 75104647 [+] |
View Full Product Details