Gene Transcripts
- Gene Symbol
- DEFA9P
- Entrez Gene ID
- 449492
- Gene Name
- defensin alpha 9, pseudogene
- Gene Aliases
- DEFAP2
- Chromosome Location
- Chr. 8: 6959289 - 6960161 on Build GRCh38
- UniGene ID
- Hs.552565
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_073408.1 |
|
2 |
231 |
|
GenBank mRNA |
AY746434.1 |
- |
2 |
231 |
|
View Full Product Details