Gene Transcripts
- Gene Symbol
- KLKP1
- Entrez Gene ID
- 606293
- Gene Name
- kallikrein pseudogene 1
- Gene Aliases
- KARMA, KLK31P, KRSP1, PsiKLK1, YKLK1
- Chromosome Location
- Chr. 19: 50882096 - 50896398 on Build GRCh38
- UniGene ID
- Hs.643424
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DQ086829.1 |
- |
1 |
351 |
|
DQ211698.1 |
- |
4 |
1667 |
|
View Full Product Details