Gene Transcripts

Gene Symbol
COX20
Entrez Gene ID
116228
Gene Name
COX20, cytochrome c oxidase assembly factor
Gene Aliases
FAM36A
Chromosome Location
Chr. 1: 244835306 - 244845063 on Build GRCh38
UniGene ID
Hs.411490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001312872.1 NP_001299801.1 2 300
GenBank mRNA BQ189361.1 - 2 160


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB097837.1 - 1 194 Chr. 1 - 244836311 - 244836899 [+]
GenBankmRNA AJ005814.1 - 1600 Chr. - - [-]

View Full Product Details