Gene Transcripts

Gene Symbol
TTLL4
Entrez Gene ID
9654
Gene Name
tubulin tyrosine ligase like 4
Gene Aliases
Chromosome Location
Chr. 2: 218710835 - 218757484 on Build GRCh38
UniGene ID
Hs.471405
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006712875.2 XP_006712938.1 20 5893
XM_017005387.1 XP_016860876.1 20 6071
XM_017005388.1 XP_016860877.1 21 5958
XM_017005389.1 XP_016860878.1 21 5977
XM_017005390.1 XP_016860879.1 20 5911
XM_017005391.1 XP_016860880.1 19 5976
XM_017005392.1 XP_016860881.1 18 6145


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK126797.1 - 1 436 Chr. 2 - 218755964 - 218759387 [+]

View Full Product Details