Gene Transcripts

Gene Symbol
C7orf49
Entrez Gene ID
78996
Gene Name
chromosome 7 open reading frame 49
Gene Aliases
MRI
Chromosome Location
Chr. 7: 135092303 - 135170826 on Build GRCh38
UniGene ID
Hs.643113
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017012589.1 XP_016868078.1 4 5816
XM_017012590.1 XP_016868079.1 4 5742
XM_017012591.1 XP_016868080.1 4 5596
XM_017012592.1 XP_016868081.1 3 5777
XM_017012593.1 XP_016868082.1 4 5646
XM_017012594.1 XP_016868083.1 4 5853


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BG773610.1 - 1 600 Chr. 7 - 135161622 - 135162300 [-]
GenBankmRNA AK056910.1 - 1 1971 Chr. 7 - 135161548 - 135163675 [-]
GenBankmRNA BG480725.1 - 1 601 Chr. 7 - 135161672 - 135162300 [-]
GenBankmRNA BE730720.1 - 581 Chr. - - [-]
GenBankmRNA BM687843.1 - 1 395 Chr. 7 - 135161513 - 135162095 [-]
GenBankmRNA BE872097.1 - 574 Chr. - - [-]

View Full Product Details