Gene Transcripts
- Gene Symbol
- C7orf49
- Entrez Gene ID
- 78996
- Gene Name
- chromosome 7 open reading frame 49
- Gene Aliases
- MRI
- Chromosome Location
- Chr. 7: 135092303 - 135170826 on Build GRCh38
- UniGene ID
- Hs.643113
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK056910.1 |
- |
1 |
2035 |
|
Chr. 7 - 135161548 - 135163675 [-] |
GenBankmRNA |
BE730720.1 |
- |
|
645 |
|
Chr. - - [-] |
GenBankmRNA |
BE873088.1 |
- |
1 |
655 |
|
Chr. 7 - 135161541 - 135162292 [-] |
GenBankmRNA |
BM687843.1 |
- |
1 |
459 |
|
Chr. 7 - 135161513 - 135162095 [-] |
View Full Product Details