Gene Transcripts
- Gene Symbol
- COL4A3BP
- Entrez Gene ID
- 10087
- Gene Name
- collagen type IV alpha 3 binding protein
- Gene Aliases
- CERT, CERTL, GPBP, MRD34, STARD11
- Chromosome Location
- Chr. 5: 75368486 - 75511981 on Build GRCh38
- UniGene ID
- Hs.270437
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125387.1 |
- |
1 |
3138 |
|
View Full Product Details