Gene Transcripts
- Gene Symbol
- ABHD3
- Entrez Gene ID
- 171586
- Gene Name
- abhydrolase domain containing 3
- Gene Aliases
- LABH3
- Chromosome Location
- Chr. 18: 21650897 - 21704805 on Build GRCh38
- UniGene ID
- Hs.397978
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CR627346.1 |
- |
|
810 |
|
View Full Product Details