Gene Transcripts
- Gene Symbol
- KATNBL1
- Entrez Gene ID
- 79768
- Gene Name
- katanin regulatory subunit B1 like 1
- Gene Aliases
- C15orf29
- Chromosome Location
- Chr. 15: 34140674 - 34210146 on Build GRCh38
- UniGene ID
- Hs.633566
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK311338.1 |
- |
6 |
1328 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK021466.1 |
- |
1 |
1038 |
|
Chr. 15 - 34145967 - 34147487 [-] |
View Full Product Details