Gene Transcripts

Gene Symbol
KATNBL1
Entrez Gene ID
79768
Gene Name
katanin regulatory subunit B1 like 1
Gene Aliases
C15orf29
Chromosome Location
Chr. 15: 34140674 - 34210146 on Build GRCh38
UniGene ID
Hs.633566
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK311338.1 - 6 1244


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK021466.1 - 1 954 Chr. 15 - 34145967 - 34147487 [-]

View Full Product Details