Gene Transcripts
- Gene Symbol
- STATH
- Entrez Gene ID
- 6779
- Gene Name
- statherin
- Gene Aliases
- STR
- Chromosome Location
- Chr. 4: 69995931 - 70002456 on Build GRCh38
- UniGene ID
- Hs.654495
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX649104.1 |
- |
1 |
2099 |
|
View Full Product Details