Gene Transcripts
- Gene Symbol
- GALNS
- Entrez Gene ID
- 2588
- Gene Name
- galactosamine (N-acetyl)-6-sulfatase
- Gene Aliases
- GALNAC6S, GAS, GalN6S, MPS4A
- Chromosome Location
- Chr. 16: 88813734 - 88856970 on Build GRCh38
- UniGene ID
- Hs.271383
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC109229.2 |
- |
1 |
925 |
|
- Gene Symbol
- LOC107987238
- Entrez Gene ID
- 107987238
- Gene Name
- proline-rich proteoglycan 2-like
- Gene Aliases
- Chromosome Location
- Chr. 16: 88823266 - 88824779 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details