Gene Transcripts

Gene Symbol
TCF12
Entrez Gene ID
6938
Gene Name
transcription factor 12
Gene Aliases
CRS3, HEB, HTF4, HsT17266, TCF-12, bHLHb20
Chromosome Location
Chr. 15: 56918090 - 57291261 on Build GRCh38
UniGene ID
Hs.511504
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK022018.1 - 1 181


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AU120541.1 - 1 181 Chr. 15 - 57072173 - 57073012 [+]

View Full Product Details