Gene Transcripts
- Gene Symbol
- TP53TG1
- Entrez Gene ID
- 11257
- Gene Name
- TP53 target 1 (non-protein coding)
- Gene Aliases
- LINC00096, NCRNA00096, P53TG1, P53TG1-D, TP53AP1
- Chromosome Location
- Chr. 7: 87325347 - 87345492 on Build GRCh38
- UniGene ID
- Hs.274329
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC068535.1 |
- |
1 |
2226 |
|
View Full Product Details