Gene Transcripts
- Gene Symbol
- SPIRE2
- Entrez Gene ID
- 84501
- Gene Name
- spire type actin nucleation factor 2
- Gene Aliases
- Spir-2
- Chromosome Location
- Chr. 16: 89817477 - 89871319 on Build GRCh38
- UniGene ID
- Hs.461786
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK130680.1 |
- |
1 |
1171 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BC035173.1 |
- |
1 |
1969 |
|
Chr. 16 - 89816769 - 89818757 [+] |
View Full Product Details