Gene Transcripts

Gene Symbol
ERBB4
Entrez Gene ID
2066
Gene Name
erb-b2 receptor tyrosine kinase 4
Gene Aliases
ALS19, HER4, p180erbB4
Chromosome Location
Chr. 2: 211375717 - 212539147 on Build GRCh38
UniGene ID
Hs.390729
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK024204.1 - 1 1818


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DR002628.1 - 1 31 Chr. 2 - 212287134 - 212287579 [-]

View Full Product Details