Gene Transcripts
- Gene Symbol
- ZMIZ2
- Entrez Gene ID
- 83637
- Gene Name
- zinc finger MIZ-type containing 2
- Gene Aliases
- NET27, TRAFIP20, ZIMP7, hZIMP7
- Chromosome Location
- Chr. 7: 44748547 - 44769881 on Build GRCh38
- UniGene ID
- Hs.600681
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK128359.1 |
- |
1 |
61 |
|
View Full Product Details