Gene Transcripts

Gene Symbol
Apbb2
Entrez Gene ID
11787
Gene Name
amyloid beta (A4) precursor protein-binding, family B, member 2
Gene Aliases
2310007D03Rik, FE65L1, Rirl1, TR2L, Zfra
Chromosome Location
Chr. 5: 66298725 - 66618817 on Build GRCm38
UniGene ID
Mm.5159
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK009197.1 - 1 299
BC022590.1 - 1 442
BC038035.1 - 1 370


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA W82585.1 - 211 Chr. - - [-]
GenBankmRNA BU581826.1 - 1 88 Chr. 5 - 66298973 - 66299364 [-]
GenBankmRNA AA199168.1 - 1 122 Chr. 5 - 66299005 - 66299446 [-]
GenBankmRNA AA397116.1 - 1 123 Chr. 5 - 66299068 - 66299446 [-]
GenBankmRNA W75718.1 - 106 Chr. - - [-]
GenBankmRNA AA571918.1 - 1 112 Chr. 5 - 66299116 - 66299435 [-]
GenBankmRNA BG086890.2 - 1 274 Chr. 5 - 66299052 - 66299596 [-]

View Full Product Details