Gene Transcripts
- Gene Symbol
- Tmem29
- Entrez Gene ID
- 382245
- Gene Name
- transmembrane protein 29
- Gene Aliases
- 2700081K05Rik, 6330540D07Rik, CXorf44-like
- Chromosome Location
- Chr. X: 150397773 - 150467331 on Build GRCm38
- UniGene ID
- Mm.277309
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK154711.1 |
- |
2 |
3529 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY526677.1 |
- |
1 |
232 |
|
Chr. X - 150416962 - 150417337 [-] |
View Full Product Details