Gene Transcripts
- Gene Symbol
- Tnfsf18
- Entrez Gene ID
- 240873
- Gene Name
- tumor necrosis factor (ligand) superfamily, member 18
- Gene Aliases
- Gitrl
- Chromosome Location
- Chr. 1: 161494657 - 161505290 on Build GRCm38
- UniGene ID
- Mm.276823
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ577580.1 |
- |
|
925 |
|
View Full Product Details