Gene Transcripts

Gene Symbol
Socs4
Entrez Gene ID
67296
Gene Name
suppressor of cytokine signaling 4
Gene Aliases
3110032M18Rik, A730004F22Rik, AI427843, AU015727, AU015859, Socs7
Chromosome Location
Chr. 14: 47277143 - 47291591 on Build GRCm38
UniGene ID
Mm.23957
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK080408.1 - 1198


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CV675449.1 - 1 836 Chr. 14 - 47294004 - 47294827 [+]
GenBankmRNA BM894612.1 - 1 299 Chr. 14 - 47294511 - 47294987 [+]
GenBankmRNA DT903349.1 - 1 649 Chr. 14 - 47294160 - 47294953 [+]
GenBankmRNA BX525705.1 - 1 178 Chr. 14 - 47294632 - 47295163 [+]
GenBankmRNA AI553361.1 - 1 177 Chr. 14 - 47294633 - 47295111 [+]
GenBankmRNA BI145262.1 - 1 462 Chr. 14 - 47294392 - 47295023 [+]
GenBankmRNA AA529335.1 - 1 177 Chr. 14 - 47294633 - 47295178 [+]

View Full Product Details