Gene Transcripts

Gene Symbol
Tmem30a
Entrez Gene ID
69981
Gene Name
transmembrane protein 30A
Gene Aliases
2010200I23Rik, AW540225, Cdc50a, D9Wsu20e
Chromosome Location
Chr. 9: 79760276 - 79794028 on Build GRCm38
UniGene ID
Mm.486324
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK078136.1 - 1 1254


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AV027935.2 - 1 321 Chr. 9 - 79768318 - 79768979 [-]
GenBankmRNA AI157016.1 - 1 333 Chr. 9 - 79768627 - 79768991 [-]
GenBankmRNA AA689995.1 - 1 103 Chr. 9 - 79768517 - 79768761 [-]
GenBankmRNA AA289871.1 - 1 374 Chr. 9 - 79768513 - 79769032 [-]
GenBankmRNA CB566185.1 - 1 425 Chr. 9 - 79768309 - 79769063 [-]
GenBankmRNA AV334659.2 - 1 545 Chr. 9 - 79768256 - 79769030 [-]
GenBankmRNA BM938637.1 - 1 531 Chr. 9 - 79768503 - 79769189 [-]
GenBankmRNA AA142508.1 - 1 263 Chr. 9 - 79768455 - 79768921 [-]
GenBankmRNA CF744147.1 - 1 469 Chr. 9 - 79768372 - 79769127 [-]

View Full Product Details