Gene Transcripts
- Gene Symbol
- Dpf2
- Entrez Gene ID
- 19708
- Gene Name
- D4, zinc and double PHD fingers family 2
- Gene Aliases
- 2210010M07Rik, BAF45D, Req, ubi-d4
- Chromosome Location
- Chr. 19: 5896516 - 5912871 on Build GRCm38
- UniGene ID
- Mm.2651
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK019051.1 |
- |
2 |
103 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY708591.1 |
- |
2 |
101 |
|
Chr. 19 - 5911826 - 5912809 [-] |
View Full Product Details