Gene Transcripts

Gene Symbol
Tcrg-V7
Entrez Gene ID
21641
Gene Name
T cell receptor gamma, variable 7
Gene Aliases
TRGV7, Vg7
Chromosome Location
Chr. 13: 19178042 - 19178492 on Build GRCm38
UniGene ID
Mm.424521
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA Z48593.1 - 1 103
Z48594.1 - 1 105


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA M71214.1 - 166 Chr. - - [-]

View Full Product Details