Gene Transcripts
- Gene Symbol
- Cet1
- Entrez Gene ID
- 12624
- Gene Name
- cerebellum expressed transcript 1
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.453061
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U03988.1 |
- |
1 |
204 |
|
View Full Product Details