Gene Transcripts
- Gene Symbol
- Lrrk1
- Entrez Gene ID
- 233328
- Gene Name
- leucine-rich repeat kinase 1
- Gene Aliases
- AW319595, C230002E15Rik, D130026O16Rik, mKIAA1790
- Chromosome Location
- Chr. 7: 66258745 - 66388341 on Build GRCm38
- UniGene ID
- Mm.33284
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK040108.1 |
- |
3 |
680 |
|
AK082077.1 |
- |
1 |
251 |
|
AK171849.1 |
- |
3 |
1506 |
|
View Full Product Details