Gene Transcripts
- Gene Symbol
- Pcgf3
- Entrez Gene ID
- 69587
- Gene Name
- polycomb group ring finger 3
- Gene Aliases
- 2310035N15Rik, AI662857, D630042K08Rik, DONG1, E430039C14, RNF3A, Rnf3
- Chromosome Location
- Chr. 5: 108461215 - 108503099 on Build GRCm38
- UniGene ID
- Mm.261218
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK009638.1 |
- |
|
786 |
|
View Full Product Details