Gene Transcripts
- Gene Symbol
- Ctso
- Entrez Gene ID
- 229445
- Gene Name
- cathepsin O
- Gene Aliases
- A330105D01Rik, A430056C07, AI118514
- Chromosome Location
- Chr. 3: 81932616 - 81956725 on Build GRCm38
- UniGene ID
- Mm.254642
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020735.1 |
- |
1 |
668 |
|
View Full Product Details