Gene Transcripts

Gene Symbol
Rian
Entrez Gene ID
75745
Gene Name
RNA imprinted and accumulated in nucleus
Gene Aliases
5530401N18Rik, C130089L09Rik, Meg8
Chromosome Location
Chr. 12: 109603945 - 109661716 on Build GRCm38
UniGene ID
Mm.293263
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF498302.1 - 2 190
EU434919.1 - 8 764


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BG694943.1 - 2 434 Chr. 12 - 109633959 - 109643464 [+]

View Full Product Details