Gene Transcripts

Gene Symbol
C17orf77
Entrez Gene ID
146723
Gene Name
chromosome 17 open reading frame 77
Gene Aliases
Chromosome Location
Chr. 17: 74584679 - 74594209 on Build GRCh38
UniGene ID
Hs.350775
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302809.1 NP_001289738.1 3 864
NM_152460.2 NP_689673.2 3 700
GenBank mRNA AK056444.1 - 3 700

View Full Product Details