Gene Transcripts
- Gene Symbol
- LOC613037
- Entrez Gene ID
- 613037
- Gene Name
- nuclear pore complex interacting protein member
- Gene Aliases
- Chromosome Location
- Chr. 16: 30222735 - 30251993 on Build GRCh38
- UniGene ID
- Hs.585733
- Species
- Homo sapiens
- Gene Symbol
- NPIPA2
- Entrez Gene ID
- 642799
- Gene Name
- nuclear pore complex interacting protein family member A2
- Gene Aliases
- NPIP
- Chromosome Location
- Chr. 16: 14745595 - 14765448 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- NPIPA7
- Entrez Gene ID
- 101059938
- Gene Name
- nuclear pore complex interacting protein family member A7
- Gene Aliases
- Chromosome Location
- Chr. 16: 16376940 - 16393972 on Build GRCh38
- UniGene ID
- Hs.634612
- Species
- Homo sapiens
- Gene Symbol
- NPIPA8
- Entrez Gene ID
- 101059953
- Gene Name
- nuclear pore complex interacting protein family member A8
- Gene Aliases
- LCR16a9
- Chromosome Location
- Chr. 16: 18317919 - 18334967 on Build GRCh38
- UniGene ID
- Hs.636569
- Species
- Homo sapiens
- Gene Symbol
- NPIPB5
- Entrez Gene ID
- 100132247
- Gene Name
- nuclear pore complex interacting protein family member B5
- Gene Aliases
- Chromosome Location
- Chr. 16: 22513523 - 22536521 on Build GRCh38
- UniGene ID
- Hs.611072
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK303572.1 |
- |
1 |
195 |
|
- Gene Symbol
- PDXDC2P
- Entrez Gene ID
- 283970
- Gene Name
- pyridoxal dependent decarboxylase domain containing 2, pseudogene
- Gene Aliases
- NPIP, PDXDC2
- Chromosome Location
- Chr. 16: 69976297 - 70065948 on Build GRCh38
- UniGene ID
- Hs.513695
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003610.1 |
|
25 |
3718 |
|
GenBank mRNA |
AL833251.1 |
- |
25 |
3718 |
|
BX647358.1 |
- |
2 |
5204 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000531894.5 |
- |
25 |
3718 |
|
Chr. 16 - 69976297 - 70065948 [-] |
View Full Product Details