Gene Transcripts

Gene Symbol
C9orf41-AS1
Entrez Gene ID
101927380
Gene Name
C9orf41 antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 9: 74952965 - 74997382 on Build GRCh38
UniGene ID
Hs.257556
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_121183.1 2 198
GenBank mRNA BC043649.2 - 2 194
BI552900.1 - 2 199


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB465744.1 - 2 187 Chr. 9 - 74952976 - 74991677 [+]
GENCODE ENST00000455609.1 - 2 195 Chr. 9 - 74952968 - 74996293 [+]

View Full Product Details