Gene Transcripts
- Gene Symbol
- HCG9
- Entrez Gene ID
- 10255
- Gene Name
- HLA complex group 9 (non-protein coding)
- Gene Aliases
- HCGIX, HCGIX4
- Chromosome Location
- Chr. 6: 29975115 - 29978403 on Build GRCh38
- UniGene ID
- Hs.656251
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028032.1 |
|
3 |
526 |
|
GenBank mRNA |
X95289.1 |
- |
3 |
521 |
|
View Full Product Details