Gene Transcripts

Gene Symbol
MUC19
Entrez Gene ID
283463
Gene Name
mucin 19, oligomeric
Gene Aliases
MUC-19
Chromosome Location
Chr. 12: 40393394 - 40570757 on Build GRCh38
UniGene ID
Hs.632712
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA CR749517.1 - 11 3721


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000474954.5 - 11 3720 Chr. 12 - 40522485 - 40534803 [+]

View Full Product Details